Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffer HF-Improved, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

044627-K2 044627-AK
AA44627-AKEA 31.59 USD InStock
AA44627-AK AA44627-K2
Buffer HF-Improved, Thermo Scientific
Buffers pH Calibration Buffers
Used for pretreatment of planar silicon devices when plating with Nickel plating solution products 44069 and 44070.

Density: 1.12
Appearance: Colorless
Odor: Acrid
Boiling Point: 103…105°
UN No.: 2817

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty