Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc oxide, NanoTek®

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Zinci oxidumZinc (II) oxide

44899-22 044899-36 044899-22 44899-36
AA44899-22EA 46.3 USD InStock
CAAA44899-22 AA44899-22 CAAA44899-36 AA44899-36
Zinc oxide, NanoTek®
Zinc oxide
NanoTek® zinc oxide, due to the small particle size, can provide UV protection while maintaining a high level of transparency in coatings, polymers, caulks, adhesives and other resin systems


Formula: ZnO
MW: 81.39 g/mol
Melting Pt: 1975 °C
Density: 5.6
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty