Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indium tin oxide ≥99.5%, NanoTek®

Supplier: Thermo Scientific Chemicals
Synonyms: ITO

44927-14 44927-22
AA44927-14EA 280.99 USD InStock
AA44927-14 AA44927-22
Indium tin oxide ≥99.5%, NanoTek®
Indium tin oxide
Applications: Indium tin oxide is one of the most widely used transparent conducting oxides due to its important properties like electrical conductivity and optical transparency, as well as the easy formation of thin film on deposition. It is also used in touch panels, electrochromic, electroluminescent and LCD displays, plasma displays, field emission displays, heat reflective coatings, energy efficient windows, gas sensors and photovoltaics.


Formula: In₂O₃·(SnO₂)x
Density: 7,16
Storage Temperature: Ambient
MDL Number: MFCD00171662
CAS Number: 50926-11-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty