Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony tin oxide ≥99.5%, NanoArc®

Supplier: Thermo Scientific Chemicals
Warning

44930.14 44930.36 44930.22
AA44930-22EA 116.99 USD InStock
AA44930-22 AA44930-14 AA44930-36
Antimony tin oxide ≥99.5%, NanoArc®
Antimony tin oxide
22-44nm APS Powder, S.A. 20-40m2/g

Applications: Moderately conductive particle. The incorporation of NanoTek™ antimony tin oxide products in coating systems, films or composites may help in reducing the buildup of static charge


Formula: Sb₂O₅:SnO₂₁₅:₈₅%
Storage Temperature: Ambient
MDL Number: MFCD00799153
UN: 1549
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty