Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ICP-MS General Memory & Interference Check Sample Mix for 6020, Specpure®, Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific
Specpure®
44935-AE 44935-AP 44935-AP 044935-AE
AA44935-APEA 557.22 USD InStock
AA44935-AP AA44935-AE CAAA44935-AE CAAA44935-AP
ICP-MS General Memory & Interference Check Sample Mix for 6020, Specpure®, Thermo Fisher Scientific
Standards Cationic Standards Multi-Element Standards
Liquid

Molecular Formula: Matrix: 2% HNO3
Physical Form: Liquid
UN No.:UN3264

Molecular Formula: Matrix: 2% HNO3
Physical Form: Liquid
UN No.:UN3264

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty