Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver conductive ink

Supplier: Thermo Scientific Chemicals
Warning

44958-06 44958-14
AA44958-14EA 302.99 USD InStock
AA44958-14 AA44958-06
Silver conductive ink
Silver conductive ink
This is a conducting screen printable thick film ink for membrane switch production.


Density: 2,3
Storage Temperature: Ambient
MDL Number: MFCD01868501
EINECS: 231-131-3
Order Now
Specification Test Results
Appearance Silver
Form Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty