Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polydimethylsiloxane emulsion

Supplier: Thermo Scientific Chemicals
Synonyms: DimethiconeSilicone oilDimethylpolysiloxanePDMS

45088-22 45088-36 45088-A4
AA45088-36EA 68.26 USD InStock
AA45088-36 AA45088-A4 AA45088-22
Polydimethylsiloxane emulsion
Polydimethylsiloxane
Liquid


Formula: (CH₃)₃SiO(Si(CH₃)₂O)n
Storage Temperature: Ambient
MDL Number: MFCD00084411
CAS Number: 63148-62-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty