Would you like to log in again?
Monel® 400 alloy, gauze woven from wire 40 mesh woven from 0.25mm (0.01in)
Supplier: Thermo Scientific Chemicals
45163-GT 45163-GT 45163-HB 045163-HB
AA45163-HBEA 93.99 USD InStock
CAAA45163-GT AA45163-HB CAAA45163-HB AA45163-GT
Monel® 400 alloy, gauze woven from wire 40 mesh woven from 0.25mm (0.01in)
Monel® 400 alloy
Typical composition of alloy: Ni 63.0% min, Cu 28.0-34.0%, C 0.3% max, Mn 2.0% max, Fe 2.5% max, S 0.024% max, Si 0.5% max
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty