Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel 61, 61Ni, Plasma Standard Solution, Specpure®, 61Ni 10 µg/ml, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Specpure®
045282-AD
AA45282-ADEA 338.99 USD InStock
AA45282-AD
Nickel 61, 61Ni, Plasma Standard Solution, Specpure®, 61Ni 10 µg/ml, Thermo Scientific
Standards Cationic Standards Nickel Standards
Molecular Formula: Matrix: 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00144901

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty