Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Germanium, Optical Window, Diameter 25.4 mm (1.0 in), Thickness 1 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

045386-KS
AA45386-KSEA 294.99 USD InStock
AA45386-KS
Germanium, Optical Window, Diameter 25.4 mm (1.0 in), Thickness 1 mm (0.04 in)
Germanium


Formula: Ge
Boiling Pt: 2830 °C (1013 hPa)
Melting Pt: 937.2 °C
Density: 5,323 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty