Would you like to log in again?
D-Glucose Dehydrogenase, Thermo Scientific
Supplier: Thermo Scientific
45402-OAT 045402-4I 045402-OAT
AA45402-OATEA 645.99 USD InStock
AA45402-OAT CAAA45402-OAT AA45402-4I
D-Glucose Dehydrogenase, Thermo Scientific
Enzymes
Uses: diagnostics and cofactor regeneration of NAD(P)H in reductive processes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty