Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cadmium selenide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cadmium (II) selenide

045456-18 045456-09
AA45456-18EA 498.99 USD InStock
AA45456-18 AA45456-09
Cadmium selenide ≥99.999% (metals basis)
Cadmium selenide


Formula: CdSe
MW: 191.37 g/mol
Melting Pt: >1350 °C
Density: 5.81
Storage Temperature: Ambient
MDL Number: MFCD00010917
CAS Number: 1306-24-7
EINECS: 215-148-3
UN: 3283
ADR: 6.1,III
Merck Index: 12,01663
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty