Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc phosphide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

045459-04 045459-02 045459-09
AA45459-09EA 933.99 USD InStock
AA45459-09 AA45459-04 AA45459-02
Zinc phosphide ≥99.999% (metals basis)
Zinc phosphide


Formula: P₂Zn₃
MW: 258.12 g/mol
Boiling Pt: 1100 °C
Melting Pt: 420 °C
Density: 4.55
Storage Temperature: Ambient
MDL Number: MFCD00049631
CAS Number: 1314-84-7
EINECS: 215-244-5
UN: 1714
ADR: 4.3,I
Merck Index: 14,10152
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty