Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver chromate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Silver(I) chromateArgentous chromate

045495-22 045495-06
AA45495-06EA 30.6 USD InStock
AA45495-06 AA45495-22
Silver chromate ≥99.9% (metals basis)
Silver chromate


Formula: Ag₂CrO₄
MW: 331.73 g/mol
Boiling Pt: 1550 °C (1013 hPa)
Density: 5.63
Storage Temperature: Ambient
MDL Number: MFCD00003402
CAS Number: 7784-01-2
EINECS: 232-043-8
UN: 1479
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty