Would you like to log in again?
Nitinol, foil, shape memory, flat annealed, pickled surface, Thickness 0.127 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
Danger
45514-GH 45514-FI 45514-FF
AA45514-FIEA 294.99 USD InStock
AA45514-FI AA45514-FF AA45514-GH
Nitinol, foil, shape memory, flat annealed, pickled surface, Thickness 0.127 mm (0.005 in)
Nitinol
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty