Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium dodecairon nonadecaoxide

Supplier: Thermo Scientific Chemicals
Warning

045552-14 045552-22 045552-06
AA45552-06EA 26.89 USD InStock
AA45552-06 AA45552-22 AA45552-14
Barium dodecairon nonadecaoxide
Barium dodecairon nonadecaoxide


Formula: BaFe₁₂O₁₉
MW: 1111.48 g/mol
Melting Pt: >1315 °C
Storage Temperature: Ambient
MDL Number: MFCD00075637
CAS Number: 12047-11-9
EINECS: 234-974-5
UN: 1564
ADR: 6.1,III
Order Now
Specification Test Results
Form Powder

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty