Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium thioglycolate trihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Calcium thioglycolate trihydrateMercaptoacetic acid calcium salt trihydrateCalcium bis(mercaptoacetate) trihydrateCalcium mercaptoacetate trihydrate

45557-18
AA45557-18EA 110.99 USD InStock
AA45557-18
Calcium thioglycolate trihydrate 98%
Calcium bis(mercaptoacetate) (Calcium thioglycolate) trihydrate


Formula: C₄H₆CaO₄S₂·3H₂O
MW: 276.35 g/mol
Melting Pt: 280…290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150707
CAS Number: 65208-41-5
EINECS: 212-402-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty