Synonyms:
Dibismuth trioxide
, Bismuth Trioxide
45582-36
45582-14
45582-22
AA45582-22EA
128.99
USD
InStock
AA45582-22
AA45582-36
AA45582-14
Bismuth(III) oxide ≥99.5%, NanoArc®
Bismuth (III) oxide
X-ray opaque particle. NanoArc™ bismuth oxide can be compounded into polymers or coating systems to increase opacity to x-rays,while the small particle size ensures minimal disruption of physical properties.