Would you like to log in again?
Magnesium Oxide Crucibles, Cylindrical with Flat Base, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
42989-KJ 045900-KS 45680-KS 42989-KJ 42989-KF 045733-KS 42991-KF 45733-KS 45854-KS 045680-KS 45900-KS 42991-KJ 042991-KF 045854-KS
AA45854-KSEA 152.99 USD InStock
AA45854-KS CAAA45733-KS CAAA45900-KS CAAA45680-KS AA42991-KF CAAA42989-KJ AA45900-KS AA45733-KS CAAA45854-KS CAAA42989-KF AA45680-KS CAAA42991-KF AA42989-KJ CAAA42991-KJ
Magnesium Oxide Crucibles, Cylindrical with Flat Base, Thermo Scientific
Crucibles
These high performance oxide ceramics display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases.
  • Cylindrical, flat bottom design
  • Crucibles provide increased working temperature characteristics
  • Manufactured of pure metal oxides without the admixtures of silicates
Oxide ceramics are also inert to oxidation and are not subject to radiation damage.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty