Would you like to log in again?
Silver, Fixing Bath, Test Strips, Thermo Scientific
Supplier: Thermo Scientific
045839-QA
AA45839-QAEA 86.59 USD InStock
AA45839-QA
Silver, Fixing Bath, Test Strips, Thermo Scientific
Environmental Test Kits
Test strips for assessing the fixing capacity of fixing baths (pH and silver (I) ion concentration) and for the detection and semi-quantitative determination of silver.
Gradation 0.5 1 2 3 5 7 10 grams/litre Ag+; 4 –5 –6 –7 –8 pH units
Caution: This product cannot be used for fixing baths with pH less than 4
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty