Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Silicone oil high temperature , colorless liquid for oil baths

Supplier: Thermo Scientific Chemicals
Synonyms: Polyphenyl-methylsiloxane

045896-36 045896-A4 045896-22
AA45896-22EA 100.31 USD InStock
AA45896-22 AA45896-36 AA45896-A4
Silicone oil high temperature , colorless liquid for oil baths
Silicone oil
Usable temperature range: 25 to 250 °C (open system) and 25 to 315 °C (closed system)

Refractive Index: 1.505

Applications: High temperature open system, closed system, circulating, closed loop, and heat transfer baths. High temperature fluids for laboratory research apparatus and instruments.


Density: 1.07 g/mL
Flash Pt: >300 °C(572 °F)
MDL Number: MFCD00132673
CAS Number: 68083-14-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty