Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lanthanum ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh

Supplier: Thermo Scientific Chemicals
Danger

045908-14 045908-06
AA45908-14EA 697.99 USD InStock
AA45908-14 AA45908-06
Lanthanum ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh
Lanthanum


Formula: La
MW: 138.91 g/mol
Boiling Pt: 3469 °C (1013 hPa)
Melting Pt: 885 °C
Density: 6.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3089
ADR: 4.1,II
Merck Index: 12,05374
Order Now
Specification Test Results
Form Packaged under argon

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty