045943-GH
045943-FI
045943-FF
45943-FF
45943-FI
45943-GH
AA45943-FFEA
58.59
USD
InStock
AA45943-FF
CAAA45943-GH
AA45943-GH
CAAA45943-FF
CAAA45943-FI
AA45943-FI
Copper ≥99.99% (metals basis), thin foil 0.008 mm (0.0003 in) thick, Oxygen-Free High Conductivity (OFHC)
Copper