Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony, plasma standard solution, Specpure®, Sb 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
47103-AP 047103-AE THERMO FISHER SCIENTIFIC
AA47103-AEEA 65.69 USD InStock
AA47103-AE AA47103-AP AA47103-AD
Antimony, plasma standard solution, Specpure®, Sb 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Antimony Standards
Antimony, plasma standard solution, Specpure|r, Sb 1000^mg/ml

Molecular Formula: Sb in 30% HCl
Physical Form: Liquid
Odor: Acidic
UN No.: 1789
MDL No.: MFCD00134030

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty