Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.8% for LC-MS

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: CyanomethaneEthanenitrileEthanoic acid nitrileMethane, cyano-Methyl cyanideMethanecarbonitrileACN

047138-K7 047138-M6 047138-K2 047138-M1
AA47138-K2EA 423.19 USD InStock
AA47138-K2 AA47138-M6 AA47138-K7 AA47138-M1
Acetonitrile ≥99.8% for LC-MS
Acetonitrile
  • Liquid

Applications: Solvent for LC-MS analysis


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 80…82 °C
Melting Pt: –48 °C… –45 °C
Density: 0.786
Flash Pt: 5 °C (41 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Assay Minimum 99.8

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty