Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gallium triiodide, ultra dry ≥99.999% (metals basis), beads -10 mesh

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Gallium (III) iodide

ampouled under argon

Ultra Dry
047148-14 047148-06 047148-03
AA47148-14EA 600.96 USD InStock
AA47148-14 AA47148-03 AA47148-06
Gallium triiodide, ultra dry ≥99.999% (metals basis), beads -10 mesh
Gallium triiodide


Formula: GaI₃
MW: 450.44 g/mol
Boiling Pt: 345 °C (subl.)
Melting Pt: 212 °C
Density: 4.15
Storage Temperature: Ambient
MDL Number: MFCD00016107
CAS Number: 13450-91-4
EINECS: 236-611-6
UN: 3260
ADR: 8,II
Order Now
Specification Test Results
Assay Minimum 99.999

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty