Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium bromide, ultra dry ≥99.999% (metals basis), powder

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Aluminum bromide

ampouled under argon

Ultra Dry
047168-06 047168-09
AA47168-06EA 333.99 USD InStock
AA47168-06 AA47168-09
Aluminium bromide, ultra dry ≥99.999% (metals basis), powder
Aluminium bromide


Formula: AlBr₃
MW: 266.69 g/mol
Boiling Pt: 265 °C
Melting Pt: 97.5 °C
Density: 3.21
Storage Temperature: Ambient
MDL Number: MFCD00003421
CAS Number: 7727-15-3
EINECS: 231-779-7
UN: 1725
ADR: 8,II
Order Now
Specification Test Results
Assay Minimum 99.999

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty