Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyltrimethylammonium hydroxide 40% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Benzyltrimethylammonium HydroxideN,N,N-Trimethyl-1-phenylmethanaminium Hydroxide

047189.AE 047189.AP
AA47189-AEEA 47.79 USD InStock
AA47189-AE AA47189-AP
Benzyltrimethylammonium hydroxide 40% (w/w) in aqueous solution
Benzyltrimethylammonium hydroxide


Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Density: 1.059
Storage Temperature: Ambient
MDL Number: MFCD00008281
CAS Number: 100-85-6
EINECS: 202-895-5
UN: 3267
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty