Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibismuth triselenide ≥99.995% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bismuth (III) selenide

047198-06 047198-03
AA47198-06EA 157.99 USD InStock
AA47198-06 AA47198-03
Dibismuth triselenide ≥99.995% (metals basis)
Dibismuth triselenide
  • -325 Mesh Powder


Formula: Bi₂Se₃
MW: 654.84 g/mol
Melting Pt: 710 °C
Density: 6.82
Storage Temperature: Ambient
MDL Number: MFCD00014200
CAS Number: 12068-69-8
EINECS: 235-104-7
UN: 3283
ADR: 6.1,III
Order Now
Specification Test Results
Assay Minimum 99.999

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty