Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium hydride ≥99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: TetrahydridozirconiumZirconium(Ⅱ)bydrideDihydridezirconium(II)

047203-30 047203-18 047203-09
AA47203-30EA 597.99 USD InStock
AA47203-30 AA47203-18 AA47203-09
Zirconium hydride ≥99%
Zirconium hydride


Formula: H₄Zr
MW: 95.256 Da
Storage Temperature: Ambient
MDL Number: MFCD00148878
CAS Number: 7704-99-6
EINECS: 231-727-3
UN: 1437
ADR: 4.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty