Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron, Plasma Standard Solution, Specpure®, Fe 1000 µg/ml, Thermo Fisher Scientific

Supplier: Thermo Fisher Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Specpure®
047206-AP 047206-AE 047206-AD
AA47206-ADEA 53.6 USD InStock
AA47206-AD AA47206-AE AA47206-AP
Iron, Plasma Standard Solution, Specpure®, Fe 1000 µg/ml, Thermo Fisher Scientific
Standards Cationic Standards Iron Standards
Iron, plasma standard solution, Specpure|r, Fe 1000^mg/ml

Molecular Formula: (Fe(NO3)3).9H2O in 5% HNO3
Physical Form: Liquid
Odor: Acidic
UN No.: 3265
MDL No.: MFCD00010999

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty