Would you like to log in again?
Tri-Foil, ArgoBraze 49LMT, 9.0w×0.4 mm thick foil strip, Thermo Scientific
Supplier: Thermo Scientific
047265-22
AA47265-22EA 113.99 USD InStock
AA47265-22
Tri-Foil, ArgoBraze 49LMT, 9.0w×0.4 mm thick foil strip, Thermo Scientific
Soldering Supplies Desoldering Tools
Trifoil 3pc strip ArgoB/Cu/ArgoB 1:2:1 clad
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty