Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyridine, anhydrous ≥99.5%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Packaged under argon.

047323-AE 047323-K2
AA47323-AEEA 173.9 USD InStock
AA47323-AE AA47323-K2
Pyridine, anhydrous ≥99.5%, AcroSeal™
Pyridine
AcroSeal™ septum bottled solvent

  • Supports optimum performance of your moisture-sensitive work


Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115 °C
Melting Pt: –42 °C
Density: 0.978
Flash Pt: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/97/790797.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty