Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrofuran ≥99.7% unstabilized, AcroSeal™, Spectrophotometric Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: OxolaneTHFCyclotetramethylene oxideTetramethylene oxide1,4-epoxy-ButaneButane alpha,delta-oxideOxacyclopentane

Packaged under argon.

047329-M1 047329-K2 047329-AK
AA47329-K2EA 122.81 USD InStock
AA47329-K2 AA47329-M1 AA47329-AK
Tetrahydrofuran ≥99.7% unstabilized, AcroSeal™, Spectrophotometric Grade
Tetrahydrofuran
AcroSeal™ septum bottled solvent.

  • Supports optimum performance of your moisture-sensitive work


Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C
Melting Pt: –108 °C
Density: 0.889
Flash Pt: –17°C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty