Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylformamide, anhydrous 99.80%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: NSC 5356Formic acid dimethylamideDimethylformamideDMFADMFFormyldimethylamineN-Formyldimethylamine

Packaged under argon.

047390-M1 047390-5Y 047390-AE 047390-K2
AA47390-M1PK 376.28 USD InStock
AA47390-M1 AA47390-5Y AA47390-AE AA47390-K2
N,N-Dimethylformamide, anhydrous 99.80%, AcroSeal™
N,N-Dimethylformamide
AcroSeal™ septum bottled solvent

  • Supports optimum performance of your moisture-sensitive work


Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C
Melting Pt: -61 °C
Density: 0.944
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (GC) ≥99.75%
Water content (Karl Fischer Titration) ≤50 ppm
Residue on evaporation ≤5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty