Synonyms:
Sodium sulphide
065122-06
65122-22
065122-22
65122-06
65122-14
065122-14
AA65122-06EA
82.98
USD
InStock
AA65122-06
CAAA65122-14
CAAA65122-06
AA65122-14
CAAA65122-22
AA65122-22
Disodium sulfide, anhydrous
Disodium sulphide
In organic chemicals, dyes rayon, leather and paper, in hydrometallurgy of gold ore, sulfiding oxidized lead and copper ores, and as an analytical reagent