Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Selenium disulfide 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Selenium sulphideSelenium(IV) sulphideSelenium sulfideSelenium(IV) sulfide

068104-18 068104-30 68104-06 68104-30 68104-18 068104-06
AA68104-30EA 374.99 USD InStock
CAAA68104-06 CAAA68104-18 CAAA68104-30 AA68104-30 AA68104-06 AA68104-18
Selenium disulfide 97%
Selenium disulphide


Formula: S₂Se
MW: 143.09 g/mol
Boiling Pt: Dec.
Melting Pt: <100 °C
Density: 0.03 g/cm³ (–65 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011227
CAS Number: 7488-56-4
EINECS: 231-303-8
UN: 2657
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty