Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium selenate ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

068111-18 68111-09 068111-09 068111-04 68111-18 68111-04
AA68111-04EA 32.49 USD InStock
CAAA68111-04 AA68111-04 CAAA68111-18 AA68111-18 CAAA68111-09 AA68111-09
Potassium selenate ≥99.5% (metals basis)
Potassium selenate


Formula: K₂O₄Se
MW: 221.15 g/mol
Density: 3,07
Storage Temperature: Ambient
MDL Number: MFCD00054125
CAS Number: 7790-59-2
EINECS: 232-214-7
UN: 2630
ADR: 6.1,I
Merck Index: 12,07836
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty