Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium hexafluorosilicate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ammonium fluorosilicate

69100-30 69100-30 69100-A7 69100-A1 069100-A7 069100-A1
AA69100-A7EA 604.99 USD InStock
CAAA69100-A7 AA69100-A7 CAAA69100-30 AA69100-30 CAAA69100-A1 AA69100-A1
Ammonium hexafluorosilicate 98%
Ammonium hexafluorosilicate
Etching glass, in soldering flux


Formula: H₈F₆N₂Si
MW: 178.15 g/mol
Melting Pt: Dec.
Density: 2,01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010887
CAS Number: 16919-19-0
EINECS: 240-968-3
UN: 2854
ADR: 6.1,III
Merck Index: 13,00529
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty