Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexafluorosilicic acid 23% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Fluosilicic acid

069109-36 69109-36
AA69109-36EA 65.19 USD InStock
CAAA69109-36 AA69109-36
Hexafluorosilicic acid 23% (w/w) in aqueous solution
Hexafluorosilicic acid


Formula: H₂SiF₆
MW: 144.09
Density: 1,223
Storage Temperature: Ambient
MDL Number: MFCD00036289
CAS Number: 16961-83-4
EINECS: 241-034-8
UN: 1778
ADR: 8,II
Order Now
Specification Test Results
Form Liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty