Synonyms:
Dibutylstannanediyl bis(2-ethylhexanoate)
, Di-n-butyltin bis(2-ethylhexanoate)
71126.14
71126-06
071126-06
71126-22
71126.22
71126-14
AA71126-06EA
38.29
USD
InStock
AA71126-06
CAAA71126-06
AA71126-14
AA71126-22
CAAA71126-14
CAAA71126-22
Dibutyltin bis(2-ethylhexanoate)
Dibutyltin bis(2-ethylhexanoate)
Soluble in mineral oil, acetone, xylene