Synonyms:
Di-n-butyltin dilaurate
071130-14
71130-14
71130-22
71130-36
071130-22
071130-36
AA71130-22EA
74.61
USD
InStock
CAAA71130-36
AA71130-22
AAA71130-14
AA71130-36
CAAA71130-22
CAAAA71130-14
Dibutyltin dilaurate ≥95%
Dibutyltin dilaurate
Soluble in benzene and ether. Insoluble in water and methanol