Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tellurate(VI) hydrate Te ≥ 45% ≥95%

Supplier: Thermo Scientific Chemicals
Warning

075106-18 75106-18 075106-09 75106-09
AA75106-18EA 259.99 USD InStock
AA75106-18 CAAA75106-09 CAAA75106-18 AA75106-09
Sodium tellurate(VI) hydrate Te ≥ 45% ≥95%
di-Sodium tellurium tetraoxide hydrate
Sodium tellurite is an intermediate in the extraction of the element, tellurium; it is a product obtained from anode slimes and is a precursor to tellurium. Solutions of sodium tellurite are used for black or blue-black coatings on iron, steel, aluminum, and copper.


Formula: Na₂O₄Te·H₂O
MW: 255.59 g/mol
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00014242
CAS Number: 10101-83-4
EINECS: 233-259-5
UN: 3284
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty