Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(IV) isopropoxide 97+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Titanium (IV) isopropoxide Titanium tetraisopropanolate

077115-22 77115-57 077115-57 077115-36 77115-22 77115-36 077115-09 77115-09
AA77115-09EA 48.69 USD InStock
CAAA77115-22 AA77115-09 CAAA77115-09 AA77115-57 CAAA77115-36 CAAA77115-57 AA77115-36 AA77115-22
Titanium(IV) isopropoxide 97+%
Titanium (IV) isopropoxide
Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids


Formula: C₁₂H₂₈O₄Ti
MW: 284.23 g/mol
Boiling Pt: 232 °C
Melting Pt: 16…20 °C
Density: 0,955
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008871
CAS Number: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3,III
Merck Index: 12,09614
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty