Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(IV) butanolate 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Titanium (IV) butoxide Titanium tetrabutanolate

77124-36 077124-36 077124-22 77124-09 077124-09 77124-22
AA77124-09EA 31.99 USD InStock
AA77124-09 CAAA77124-09 CAAA77124-36 AA77124-22 CAAA77124-22 AA77124-36
Titanium(IV) butanolate 99+%
Titanium(IV) butanolate
  • Liquid


Formula: C₁₆H₃₆O₄Ti
MW: 340.34 g/mol
Boiling Pt: 310…314 °C
Melting Pt: –55 °C
Density: 0.998
Flash Pt: 41 °C (170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009433
CAS Number: 5593-70-4
EINECS: 227-006-8
UN: 2924
ADR: 3,III
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty