Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopentadienylvanadium tetracarbonyl ≥97%

Supplier: Thermo Scientific Chemicals
Danger

81123-04 81123.02 81123.04 81123-02
AA81123-04EA 451.99 USD InStock
AA81123-04 CAAA81123-04 AA81123-02 CAAA81123-02
Cyclopentadienylvanadium tetracarbonyl ≥97%
Cyclopentadienylvanadium tetracarbonyl


Formula: C₉H₅O₄V
Boiling Pt: 80…100 °C (0.1 mmHg) (subl.)
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00014259
CAS Number: 12108-04-2
EINECS: 235-163-9
UN: 3466
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty