Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylmagnesium bromide 3 M in ether

Supplier: Thermo Scientific Chemicals
Danger

87291-JU 087291.JU 087291.JT 87291-JT
AA87291-JTEA 69.21 USD InStock
CAAA87291-JT CAAA87291-JU AA87291-JT AA87291-JU
Ethylmagnesium bromide 3 M in ether
Ethylmagnesium bromide
Applications: Grignard reagent


Formula: C₂H₅BrMg
Boiling Pt: 34.6 °C
Melting Pt: –116.3 °C
Density: 1,020
Storage Temperature: Ambient
MDL Number: MFCD00000043
CAS Number: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty