Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tungstate dihydrate ≥95%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium wolframate dihydrate

087371-30 087371-A1 87371-A1 87371-30
AA87371-A1EA 271 USD InStock
CAAA87371-30 AA87371-A1 CAAA87371-A1 AA87371-30
Sodium tungstate dihydrate ≥95%
Sodium wolframate dihydrate


Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Melting Pt: 100 °C (-2H₂O)
Density: 3,250
MDL Number: MFCD00149190
CAS Number: 10213-10-2
Merck Index: 12,08847
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty