Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogensulfide, anhydrous

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium hydrogen sulphide

87389-04 87389-09 087389-04 087389-09
AA87389-09EA 816.99 USD InStock
AA87389-09 AA87389-04 CAAA87389-09 CAAA87389-04
Sodium hydrogensulfide, anhydrous
Sodium hydrogensulphide
In paper pulping, in dyestuffs processing, in rayon and cellophane desulfurizing, in unhairing hides and as a bleaching reagent


Formula: HNaS
MW: 56.06 g/mol
Melting Pt: 350 °C
Density: 1.79
Storage Temperature: Ambient
MDL Number: MFCD00011124
CAS Number: 16721-80-5
EINECS: 240-778-0
UN: 2318
ADR: 4.2,II
Merck Index: 12,08730
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty