087758-14
87758-22
87758-14
087758-06
87758-36
87758.22
87758-06
87758.36
AA87758-06EA
31.99
USD
InStock
AA87758-06
AA87758-14
AA87758-36
CAAA87758-06
CAAA87758-22
CAAA87758-14
CAAA87758-36
AA87758-22
Cobalt(II) oxalate dihydrate, Reagent Grade
Cobalt (II) oxalate dihydrate
Applications: Stabilizer for HCN, temperature indicator
Soluble in aqueous ammonia. Insoluble in water. Slightly soluble in acids